Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008992.2 | NP_033018.2 | 10 - 11 | 1169 | 90 | |
GenBank mRNA | AJ001166.1 | - | 10 - 11 | 1073 | 90 | |
AK028420.1 | - | 10 - 11 | 1207 | 90 | ||
BC050102.1 | - | 10 - 11 | 1277 | 30012556 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |