Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mab21l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172295.4 | NP_758499.3 | 3 - 4 | 707 | 74 | |
GenBank mRNA | AK161057.1 | - | 3 - 4 | 624 | 74 | |
AK166928.1 | - | 457 | 74 | |||
AK172389.1 | - | 3 - 4 | 656 | 74 | ||
BC037703.1 | - | 3 - 4 | 644 | 4166596 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |