Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199198.2 | NP_954668.2 | 13 - 14 | 1580 | 88 | |
NR_028447.1 | 13 - 14 | 1591 | 88 | |||
NR_028448.1 | 12 - 13 | 1511 | 88 | |||
NR_028449.1 | 13 - 14 | 1591 | 88 | |||
GenBank mRNA | AK040235.1 | - | 13 - 14 | 1543 | 88 | |
AK045028.1 | - | 12 - 13 | 1791 | 88 | ||
AK154045.1 | - | 12 - 13 | 1475 | 88 | ||
BC013700.1 | - | 13 - 14 | 1544 | 88 | ||
BC064018.1 | - | 13 - 14 | 1507 | 3980699 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |