Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018803.2 | NP_061273.1 | 1 - 2 | 83 | ||
GenBank mRNA | AB026807.1 | - | 1 - 2 | 148 | 83 | |
BC125632.1 | - | 1 - 2 | 40130147 | 83 | ||
BC125634.1 | - | 1 - 2 | 40130149 | 83 | ||
CJ093642.1 | - | 1 - 2 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |