Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011161.5 | NP_035291.4 | 9 - 10 | 851 | 64 | |
GenBank mRNA | AF135185.1 | - | 9 - 10 | 831 | 64 | |
AK144846.1 | - | 9 - 10 | 824 | 64 | ||
AK161190.1 | - | 7 - 8 | 1026 | 64 | ||
AK161562.1 | - | 9 - 10 | 803 | 64 | ||
BC057211.1 | - | 9 - 10 | 787 | 64 | ||
BC092526.1 | - | 9 - 10 | 851 | 6593321 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |