Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300008074.mm.1 |
NM_028892.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008074.mm.1 |
NM_028892.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008074.mm.1 |
TC0300000992.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300008070.mm.1 |
NM_028892.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008070.mm.1 |
NM_028892.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008070.mm.1 |
TC0300000992.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300008073.mm.1 |
NM_028892.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008073.mm.1 |
NM_028892.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008073.mm.1 |
TC0300000992.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300008071.mm.1 |
NM_028892.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008071.mm.1 |
NM_028892.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008071.mm.1 |
TC0300000992.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300008072.mm.1 |
NM_028892.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008072.mm.1 |
NM_028892.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008072.mm.1 |
TC0300000992.mm.1 |
MTA TC |