Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027388.2 | NP_081664.2 | 1 - 2 | 476 | 87 | |
GenBank mRNA | AK081337.1 | - | 1 - 2 | 477 | 87 | |
BB647852.1 | - | 1 - 2 | 477 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |