Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300008325.mm.1 |
NM_010330.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008325.mm.1 |
NM_010330.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008325.mm.1 |
TC1300001326.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300008326.mm.1 |
NM_010330.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008326.mm.1 |
NM_010330.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008326.mm.1 |
TC1300001326.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300008329.mm.1 |
NM_010330.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008329.mm.1 |
NM_010330.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008329.mm.1 |
TC1300001326.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300008330.mm.1 |
NM_010330.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008330.mm.1 |
NM_010330.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008330.mm.1 |
TC1300001326.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300008327.mm.1 |
NM_010330.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008327.mm.1 |
NM_010330.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008327.mm.1 |
TC1300001326.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300008324.mm.1 |
NM_010330.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008324.mm.1 |
NM_010330.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008324.mm.1 |
TC1300001326.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300008328.mm.1 |
NM_010330.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008328.mm.1 |
NM_010330.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008328.mm.1 |
TC1300001326.mm.1 |
MTA TC |