Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010937.2 | NP_035067.2 | 1 - 2 | 149 | 104 | |
GenBank mRNA | AK011704.1 | - | 1 - 2 | 96 | 104 | |
AK150357.1 | - | 1 - 2 | 88 | 104 | ||
AK165960.1 | - | 1 - 2 | 93 | 104 | ||
BC051443.1 | - | 1 - 2 | 72 | 30061371 | 104 | |
BY321840.1 | - | 1 - 2 | 149 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |