Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008979.2 | NP_033005.1 | 19 - 20 | 2390 | 102 | |
NR_104070.1 | 21 - 22 | 2407 | 102 | |||
GenBank mRNA | AK153812.1 | - | 21 - 22 | 2408 | 102 | |
M90388.1 | - | 19 - 20 | 2375 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mus musculus Probe spans exons |