Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap4b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163552.1 | NP_001157024.1 | 8 - 9 | 1519 | 73 | |
NM_001163553.1 | NP_001157025.1 | 6 - 7 | 1648 | 73 | ||
NM_026193.2 | NP_080469.2 | 7 - 8 | 1732 | 73 | ||
GenBank mRNA | AK007733.1 | - | 7 - 8 | 1621 | 73 | |
AK165570.1 | - | 6 - 7 | 1568 | 73 | ||
BC054092.1 | - | 8 - 9 | 1530 | 30006597 | 73 | |
BC056200.2 | - | 7 - 8 | 1620 | 30015292 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |