Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrig2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025067.1 | NP_001020238.1 | 1 - 2 | 406 | 58 | |
GenBank mRNA | AK036116.1 | - | 1 - 2 | 407 | 58 | |
AK155089.1 | - | 1 - 2 | 469 | 58 | ||
AK158022.1 | - | 2 - 3 | 423 | 58 | ||
BC094228.1 | - | 1 - 2 | 343 | 30362610 | 58 | |
BC138422.1 | - | 1 - 2 | 317 | 8861442 | 58 | |
CD355189.1 | - | 2 - 3 | 261 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |