Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hipk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301304.1 | NP_001288233.1 | 8 - 9 | 2156 | 88 | |
NM_001301306.1 | NP_001288235.1 | 8 - 9 | 2266 | 88 | ||
NM_010432.2 | NP_034562.2 | 8 - 9 | 2266 | 88 | ||
GenBank mRNA | AF071070.1 | - | 8 - 9 | 2156 | 88 | |
AF077658.1 | - | 8 - 9 | 2170 | 88 | ||
AF170303.1 | - | 8 - 9 | 2066 | 88 | ||
AF170304.1 | - | 8 - 9 | 2066 | 88 | ||
AK122333.1 | - | 1 - 2 | 163 | 88 | ||
AK134408.1 | - | 8 - 9 | 2267 | 88 | ||
BC145697.1 | - | 8 - 9 | 2019 | 40131000 | 88 | |
BC145699.1 | - | 8 - 9 | 2019 | 40131002 | 88 | |
DQ279086.1 | - | 2 - 3 | 350 | 88 | ||
DQ279087.1 | - | 2 - 3 | 352 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |