Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172958.3 | NP_766546.1 | 11 - 12 | 1206 | 63 | |
GenBank mRNA | AK030139.1 | - | 11 - 12 | 1273 | 63 | |
AK050099.1 | - | 11 - 12 | 1199 | 63 | ||
AK050236.1 | - | 11 - 12 | 1169 | 63 | ||
AK076161.1 | - | 3 - 4 | 286 | 63 | ||
AK122539.1 | - | 11 - 12 | 1264 | 63 | ||
BC042573.1 | - | 11 - 12 | 1195 | 63 | ||
BC054352.1 | - | 11 - 12 | 1206 | 3661020 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |