Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011767.2 | NP_035897.2 | 2 - 3 | 198 | 129 | |
GenBank mRNA | AF071059.1 | - | 2 - 3 | 196 | 129 | |
BC038599.1 | - | 2 - 3 | 202 | 129 | ||
BC058570.1 | - | 2 - 3 | 198 | 129 | ||
BC094220.1 | - | 1 - 2 | 86 | 129 | ||
BC141139.1 | - | 2 - 3 | 182 | 9055751 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |