Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpped1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172610.3 | NP_766198.1 | 4 - 5 | 798 | 58 | |
GenBank mRNA | AK048421.1 | - | 4 - 5 | 799 | 58 | |
AK158740.1 | - | 4 - 5 | 817 | 58 | ||
AK165755.1 | - | 4 - 5 | 759 | 58 | ||
BC057550.1 | - | 3 - 4 | 723 | 6810744 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |