Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cuedc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172099.1 | NP_001165570.1 | 3 - 4 | 866 | 71 | |
NM_198013.3 | NP_932130.2 | 4 - 5 | 1064 | 71 | ||
GenBank mRNA | AK049250.1 | - | 4 - 5 | 1028 | 71 | |
BC024476.1 | - | 4 - 5 | 913 | 4976954 | 71 | |
BC025434.1 | - | 4 - 5 | 918 | 71 | ||
BC029708.1 | - | 4 - 5 | 997 | 5041450 | 71 | |
BC131989.1 | - | 3 - 4 | 837 | 40130266 | 71 | |
BC131991.1 | - | 3 - 4 | 837 | 40130268 | 71 | |
BC145206.1 | - | 3 - 4 | 837 | 9053757 | 71 | |
CK332869.1 | - | 3 - 4 | 310 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |