Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc64b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153784.2 | NP_722479.1 | 4 - 5 | 657 | 54 | |
GenBank mRNA | AK036793.1 | - | 4 - 5 | 672 | 54 | |
AK133729.1 | - | 4 - 5 | 674 | 54 | ||
BC038613.1 | - | 4 - 5 | 683 | 4458084 | 54 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |