Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2310002D06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045490.1 | 2 - 3 | 119 | 70 | ||
GenBank mRNA | AK009088.1 | - | 2 - 3 | 120 | 70 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 26777 |
Gm26777 | Anti-sense | Chr. 12 - 80509945 - 80518157 on NCBI Build GRCm38 | ||
Downstream | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 16 | Galnt16 |
5730405L21Rik, AI415388, Galntl1, mpp-GalNAc-T16 |
359.0 | Sense | Chr. 12 - 80518313 - 80603896 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons |