Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013026.2 | NP_001013044.2 | 19 - 20 | 2980 | 98 | |
GenBank mRNA | AK014607.1 | - | 19 - 20 | 2968 | 98 | |
AK087352.1 | - | 19 - 20 | 2980 | 98 | ||
BC087733.1 | - | 19 - 20 | 2987 | 30434981 | 98 | |
BC096625.1 | - | 19 - 20 | 2978 | 30109731 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |