Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013486.2 | NP_038514.1 | 1 - 2 | 114 | 66 | |
GenBank mRNA | AF065906.1 | - | 1 - 2 | 111 | 66 | |
AF065907.1 | - | 1 - 2 | 111 | 66 | ||
AF065908.1 | - | 1 - 2 | 111 | 66 | ||
AF065909.1 | - | 1 - 2 | 111 | 66 | ||
AF306543.1 | - | 1 - 2 | 111 | 66 | ||
AK138728.1 | - | 1 - 2 | 96 | 66 | ||
BC053731.1 | - | 1 - 2 | 97 | 30077134 | 66 | |
M18934.1 | - | 1 - 2 | 101 | 66 | ||
X06143.1 | - | 1 - 2 | 69 | 66 | ||
Y00023.1 | - | 1 - 2 | 114 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |