Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009833.1 | NP_033963.1 | 2 - 3 | 239 | 144 | |
GenBank mRNA | AF087662.1 | - | 2 - 3 | 239 | 144 | |
AF095640.1 | - | 2 - 3 | 239 | 144 | ||
AF109179.1 | - | 2 - 3 | 239 | 144 | ||
AF113951.1 | - | 2 - 3 | 239 | 144 | ||
AK083037.1 | - | 2 - 3 | 283 | 144 | ||
AK133168.1 | - | 939 | 144 | |||
BC131685.1 | - | 2 - 3 | 239 | 40111509 | 144 | |
BC131686.1 | - | 2 - 3 | 239 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |