Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025287.2 | NP_079563.2 | 2 - 3 | 118 | 139 | |
GenBank mRNA | AK004669.1 | - | 2 - 3 | 88 | 139 | |
AK028201.1 | - | 1 - 2 | 254 | 139 | ||
AK030746.1 | - | 1 - 2 | 305 | 139 | ||
AK132383.1 | - | 1 - 2 | 294 | 139 | ||
AK159482.1 | - | 1 - 2 | 271 | 139 | ||
AK160858.1 | - | 1 - 2 | 214 | 140 | ||
AK166581.1 | - | 1 - 2 | 228 | 139 | ||
AK168185.1 | - | 1 - 2 | 240 | 139 | ||
AK168546.1 | - | 1 - 2 | 307 | 139 | ||
BC043131.1 | - | 1 - 2 | 137 | 6811159 | 139 | |
BC045205.1 | - | 2 - 3 | 88 | 4988521 | 139 | |
BC064809.1 | - | 1 - 2 | 242 | 139 | ||
BY303667.1 | - | 2 - 3 | 123 | 140 | ||
BY726358.1 | - | 2 - 3 | 119 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |