Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025287.2 | NP_079563.2 | 8 - 9 | 905 | 81 | |
GenBank mRNA | AB071989.1 | - | 6 - 7 | 714 | 81 | |
AK004669.1 | - | 8 - 9 | 875 | 81 | ||
AK028201.1 | - | 7 - 8 | 1041 | 81 | ||
AK030746.1 | - | 7 - 8 | 1092 | 81 | ||
AK132383.1 | - | 7 - 8 | 1081 | 81 | ||
AK159482.1 | - | 7 - 8 | 1058 | 81 | ||
AK160858.1 | - | 7 - 8 | 1003 | 81 | ||
AK166581.1 | - | 7 - 8 | 1015 | 81 | ||
AK168185.1 | - | 7 - 8 | 1027 | 81 | ||
AK168546.1 | - | 7 - 8 | 1094 | 81 | ||
AY538613.1 | - | 6 - 7 | 717 | 81 | ||
BC043131.1 | - | 7 - 8 | 924 | 6811159 | 81 | |
BC045205.1 | - | 8 - 9 | 875 | 4988521 | 81 | |
BC064809.1 | - | 7 - 8 | 1029 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |