Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nle1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145431.2 | NP_663406.2 | 10 - 11 | 1253 | 105 | |
GenBank mRNA | AK141230.1 | - | 10 - 11 | 1228 | 105 | |
AK170853.1 | - | 10 - 11 | 1226 | 105 | ||
BC018399.1 | - | 10 - 11 | 1244 | 3491925 | 105 | |
BC035421.1 | - | 10 - 11 | 1244 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |