Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rffl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007465.3 | NP_001007466.1 | 4 - 5 | 882 | 61 | |
NM_001164569.1 | NP_001158041.1 | 5 - 6 | 1026 | 61 | ||
NM_001164570.1 | NP_001158042.1 | 5 - 6 | 968 | 61 | ||
GenBank mRNA | AK128983.1 | - | 5 - 6 | 969 | 61 | |
AK150213.1 | - | 4 - 5 | 815 | 61 | ||
AK150541.1 | - | 5 - 6 | 988 | 61 | ||
AK159442.1 | - | 4 - 5 | 841 | 61 | ||
AK169988.1 | - | 4 - 5 | 883 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |