Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013375.1 | NP_001013393.1 | 8 - 9 | 1208 | 78 | |
GenBank mRNA | AK078033.1 | - | 3 - 4 | 344 | 78 | |
AK154637.1 | - | 8 - 9 | 1148 | 78 | ||
AK167248.1 | - | 8 - 9 | 1207 | 78 | ||
BC119621.1 | - | 8 - 9 | 1181 | 40062238 | 78 | |
BC119622.1 | - | 8 - 9 | 1181 | 40062245 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |