Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015830.1 | NP_056645.1 | 1 - 2 | 1475 | 69 | |
GenBank mRNA | AK143711.1 | - | 3 - 4 | 2114 | 69 | |
AK170543.1 | - | 2 - 3 | 1709 | 69 | ||
BC058094.1 | - | 2 - 3 | 1733 | 6824892 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |