Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Decr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011933.2 | NP_036063.1 | 7 - 8 | 781 | 123 | |
GenBank mRNA | AF155575.1 | - | 7 - 8 | 781 | 123 | |
AK137212.1 | - | 7 - 8 | 712 | 123 | ||
AK149270.1 | - | 7 - 8 | 906 | 123 | ||
AK162961.1 | - | 7 - 8 | 917 | 123 | ||
BC021865.1 | - | 7 - 8 | 753 | 5123899 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |