Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nos2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010927.3 | NP_035057.1 | 20 - 21 | 2595 | 60 | |
GenBank mRNA | AF065919.1 | - | 20 - 21 | 2592 | 60 | |
AF065920.1 | - | 20 - 21 | 2592 | 60 | ||
AF065921.2 | - | 20 - 21 | 2592 | 60 | ||
AF065922.2 | - | 19 - 20 | 2465 | 60 | ||
AF065923.2 | - | 19 - 20 | 2465 | 60 | ||
AY090567.1 | - | 19 - 20 | 2415 | 60 | ||
BC062378.1 | - | 20 - 21 | 2604 | 4978648 | 60 | |
M84373.1 | - | 20 - 21 | 2596 | 60 | ||
M87039.1 | - | 20 - 21 | 2667 | 60 | ||
M92649.1 | - | 20 - 21 | 2603 | 60 | ||
U43428.1 | - | 20 - 21 | 2483 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |