Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008702.3 | NP_032728.3 | 7 - 8 | 2309 | 86 | |
GenBank mRNA | AF036332.1 | - | 7 - 8 | 1436 | 86 | |
AK078118.1 | - | 7 - 8 | 1418 | 86 | ||
BC057667.1 | - | 7 - 8 | 1394 | 4235238 | 86 | |
BC058652.1 | - | 7 - 8 | 2257 | 6406851 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |