Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxct1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024188.6 | NP_077150.1 | 7 - 8 | 1068 | 76 | |
GenBank mRNA | AB085609.1 | - | 7 - 8 | 810 | 76 | |
AK009027.1 | - | 7 - 8 | 848 | 76 | ||
AK010790.1 | - | 7 - 8 | 811 | 76 | ||
AK011354.1 | - | 7 - 8 | 748 | 76 | ||
AK078086.1 | - | 7 - 8 | 1068 | 76 | ||
AK146158.1 | - | 7 - 8 | 826 | 76 | ||
AK146342.1 | - | 7 - 8 | 813 | 76 | ||
AK151662.1 | - | 7 - 8 | 807 | 76 | ||
AK151695.1 | - | 7 - 8 | 798 | 76 | ||
AK167681.1 | - | 7 - 8 | 898 | 76 | ||
BC003422.1 | - | 7 - 8 | 772 | 2598824 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |