Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xrcc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010247.2 | NP_034377.2 | 2 - 3 | 199 | 131 | |
GenBank mRNA | AB010282.1 | - | 2 - 3 | 140 | 131 | |
AF483500.1 | - | 1 - 2 | 79 | 131 | ||
AF483501.1 | - | 1 - 2 | 79 | 131 | ||
AK143570.1 | - | 2 - 3 | 174 | 131 | ||
AK146392.1 | - | 2 - 3 | 118 | 131 | ||
AK146394.1 | - | 2 - 3 | 118 | 131 | ||
AK146396.1 | - | 2 - 3 | 166 | 131 | ||
BC031422.1 | - | 2 - 3 | 148 | 3669715 | 131 | |
BC051085.1 | - | 2 - 3 | 1377 | 131 | ||
BY292222.1 | - | 2 - 3 | 201 | 131 | ||
Z14157.1 | - | 337 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |