Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipo9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153774.1 | NP_722469.1 | 10 - 11 | 1182 | 87 | |
GenBank mRNA | AF273672.1 | - | 10 - 11 | 1127 | 87 | |
AF273673.1 | - | 10 - 11 | 1127 | 87 | ||
AJ309238.1 | - | 10 - 11 | 1121 | 87 | ||
AK036687.1 | - | 10 - 11 | 1132 | 87 | ||
BC056469.1 | - | 10 - 11 | 1149 | 87 | ||
BC079546.1 | - | 11 - 12 | 1293 | 87 | ||
BC098508.1 | - | 10 - 11 | 1132 | 30015513 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |