Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipo9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153774.1 | NP_722469.1 | 5 - 6 | 664 | 79 | |
GenBank mRNA | AF273673.1 | - | 5 - 6 | 609 | 79 | |
AJ309238.1 | - | 5 - 6 | 603 | 79 | ||
AK003047.1 | - | 5 - 6 | 79 | |||
AK036687.1 | - | 5 - 6 | 614 | 79 | ||
AK173127.1 | - | 5 - 6 | 604 | 79 | ||
BC056469.1 | - | 5 - 6 | 631 | 79 | ||
BC079546.1 | - | 6 - 7 | 775 | 79 | ||
BC098508.1 | - | 5 - 6 | 614 | 30015513 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |