Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vgll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133251.1 | NP_573514.1 | 1 - 2 | 48 | 74 | |
GenBank mRNA | AB036708.1 | - | 1 - 2 | 48 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |