Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100011954.mm.1 |
NM_080844.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011954.mm.1 |
NM_080844.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011954.mm.1 |
TC0100001462.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011961.mm.1 |
NM_080844.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011961.mm.1 |
NM_080844.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011961.mm.1 |
TC0100001462.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011959.mm.1 |
NM_080844.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011959.mm.1 |
NM_080844.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011959.mm.1 |
TC0100001462.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011955.mm.1 |
NM_080844.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011955.mm.1 |
NM_080844.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011955.mm.1 |
TC0100001462.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011960.mm.1 |
NM_080844.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011960.mm.1 |
NM_080844.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011960.mm.1 |
TC0100001462.mm.1 |
MTA TC |