Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sri | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080974.2 | NP_001074443.1 | 3 - 4 | 255 | 63 | |
NM_025618.3 | NP_079894.2 | 3 - 4 | 295 | 63 | ||
GenBank mRNA | AK008404.1 | - | 3 - 4 | 247 | 63 | |
AK008783.1 | - | 3 - 4 | 296 | 63 | ||
AK008970.1 | - | 3 - 4 | 244 | 63 | ||
AK134747.1 | - | 3 - 4 | 257 | 63 | ||
AK146071.1 | - | 3 - 4 | 252 | 63 | ||
AK146911.1 | - | 3 - 4 | 236 | 63 | ||
AK168572.1 | - | 3 - 4 | 236 | 63 | ||
AK168792.1 | - | 3 - 4 | 236 | 63 | ||
AK168900.1 | - | 3 - 4 | 227 | 63 | ||
BC065790.1 | - | 3 - 4 | 222 | 948616 | 63 | |
BY708689.1 | - | 3 - 4 | 293 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |