Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008715.2 | NP_032741.2 | 11 - 12 | 1843 | 123 | |
GenBank mRNA | AK046055.1 | - | 4 - 5 | 458 | 123 | |
BC059263.1 | - | 11 - 12 | 1843 | 6409580 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |