Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177381.3 | NP_796355.2 | 21 - 22 | 2407 | 74 | |
GenBank mRNA | BC038030.1 | - | 22 - 23 | 2418 | 4020725 | 74 |
CF749874.1 | - | 3 - 4 | 292 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |