Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033488.2 | NP_001028660.2 | 10 - 11 | 1054 | 70 | |
GenBank mRNA | AK156138.1 | - | 11 - 12 | 1206 | 70 | |
AK157892.1 | - | 9 - 10 | 1881 | 70 | ||
AK171660.1 | - | 7 - 8 | 717 | 70 | ||
AK172423.1 | - | 10 - 11 | 1055 | 70 | ||
BC145584.1 | - | 10 - 11 | 1067 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |