Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033488.2 | NP_001028660.2 | 1 - 2 | 87 | 63 | |
GenBank mRNA | AK133061.1 | - | 1 - 2 | 88 | 63 | |
AK157892.1 | - | 1 - 2 | 74 | 63 | ||
AK162620.1 | - | 1 - 2 | 86 | 63 | ||
AK171660.1 | - | 1 - 2 | 84 | 63 | ||
AK172423.1 | - | 1 - 2 | 88 | 63 | ||
BC145584.1 | - | 1 - 2 | 86 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |