Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd13c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013806.1 | NP_001013828.1 | 9 - 10 | 1276 | 116 | |
GenBank mRNA | AK135906.1 | - | 10 - 11 | 1012 | 116 | |
AK157542.1 | - | 1458 | 116 | |||
BC065088.1 | - | 8 - 9 | 3470 | 116 | ||
BC138991.1 | - | 9 - 10 | 1284 | 8862013 | 116 | |
BC138992.1 | - | 10 - 11 | 1263 | 8862014 | 116 | |
CF736962.1 | - | 2 - 3 | 272 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |