Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011785.3 | NP_035915.3 | 9 - 10 | 992 | 81 | |
GenBank mRNA | AF124142.1 | - | 9 - 10 | 982 | 81 | |
AK052953.1 | - | 1 - 2 | 104 | 81 | ||
BC066861.1 | - | 9 - 10 | 992 | 30089997 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |