Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fsd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172904.2 | NP_766492.2 | 9 - 10 | 1634 | 78 | |
GenBank mRNA | AK036674.1 | - | 1600 | 78 | ||
AK052866.1 | - | 3 - 4 | 434 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |