Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap3b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021492.3 | NP_067467.2 | 23 - 24 | 3166 | 63 | |
GenBank mRNA | AB030202.1 | - | 9 - 10 | 1040 | 63 | |
AY528675.1 | - | 23 - 24 | 2947 | 63 | ||
BC030306.1 | - | 6 - 7 | 830 | 63 | ||
BC139378.1 | - | 23 - 24 | 2914 | 8862400 | 63 | |
BC139379.1 | - | 23 - 24 | 2914 | 8862401 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |