Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081123.1 | NP_001074592.1 | 7 - 8 | 1075 | 95 | |
GenBank mRNA | AB073967.1 | - | 7 - 8 | 942 | 95 | |
AK048167.1 | - | 7 - 8 | 964 | 95 | ||
BC090672.1 | - | 7 - 8 | 1046 | 30941825 | 95 | |
BC145645.1 | - | 7 - 8 | 1041 | 40130948 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |