Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081363.2 | NP_001074832.2 | 16 - 17 | 8757 | 73 | |
GenBank mRNA | AF194970.1 | - | 5 - 6 | 7042 | 73 | |
AK078350.1 | - | 3 - 4 | 768 | 73 | ||
BC023896.1 | - | 4 - 5 | 1509 | 73 | ||
DQ642022.1 | - | 15 - 16 | 8614 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |