Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024221.3 | NP_077183.1 | 1 - 2 | 100 | 93 | |
GenBank mRNA | AK011810.1 | - | 64 | 93 | ||
AK014918.1 | - | 1 - 2 | 101 | 93 | ||
AK166631.1 | - | 1 - 2 | 102 | 93 | ||
BC019512.1 | - | 1 - 2 | 73 | 4210204 | 93 | |
CJ159324.1 | - | 1 - 2 | 104 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons |