Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161667.1 | NP_001155139.1 | 9 - 10 | 1299 | 80 | |
NM_053115.2 | NP_444345.2 | 9 - 10 | 1424 | 80 | ||
GenBank mRNA | AJ238492.1 | - | 9 - 10 | 1341 | 80 | |
AK144243.1 | - | 9 - 10 | 1293 | 80 | ||
BC021339.1 | - | 9 - 10 | 1303 | 5052291 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |